Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 64470793 - 64693058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293274.1 NP_001280203.1 14 - 15 2510 114
NM_020925.2 NP_065976.2 15 - 16 2231 114
XM_011541862.1 XP_011540164.1 14 - 15 2506 114
XM_011541863.1 XP_011540165.1 15 - 16 2655 114
GenBank mRNA AB046793.1 - 13 - 14 2013 114
AK022571.1 - 2 - 3 329 114
AK024256.1 - 5 - 6 656 114
AL833000.1 - 5 - 6 680 114
BC039301.1 - 15 - 16 2217 114

View Full Product Details