Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 64470793 - 64693058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293274.1 NP_001280203.1 16 - 17 2744 126
NM_020925.2 NP_065976.2 17 - 18 2465 126
XM_011541862.1 XP_011540164.1 16 - 17 2740 126
XM_011541863.1 XP_011540165.1 17 - 18 2889 126
GenBank mRNA AB046793.1 - 15 - 16 2247 126
AK022571.1 - 4 - 5 563 126
AK024256.1 - 7 - 8 890 126
AK299361.1 - 3 - 4 308 126
AL833000.1 - 7 - 8 914 126
BC039301.1 - 17 - 18 2451 126
BX537603.1 - 16 - 17 2743 126

View Full Product Details