The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020925.2 | NP_065976.2 | 2 - 3 | 214 | 99 |
GenBank mRNA | CX163220.1 | - | 2 - 3 | 205 | 99 |