The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033119.4 | NP_149110.1 | 2 - 3 | 284 | 94 |
XM_011523399.2 | XP_011521701.1 | 2 - 3 | 218 | 94 | |
XM_017023804.1 | XP_016879293.1 | 2 - 3 | 218 | 94 | |
GenBank mRNA | AB062886.1 | - | 2 - 3 | 65 | 94 |
AF358135.1 | - | 2 - 3 | 67 | 94 | |
AK314930.1 | - | 2 - 3 | 218 | 94 | |
AY061883.1 | - | 2 - 3 | 218 | 94 | |
BC051288.1 | - | 2 - 3 | 284 | 94 |