Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKD1
Entrez Gene ID
85407
Gene Name
naked cuticle homolog 1
Gene Aliases
Naked1
Chromosome Location
Chr.16: 50548330 - 50649249 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033119.4 NP_149110.1 6 - 7 690 59
XM_011523399.2 XP_011521701.1 9 - 10 1001 59
XM_011523400.2 XP_011521702.1 4 - 5 292 59
XM_017023804.1 XP_016879293.1 8 - 9 2220 59
GenBank mRNA AB062886.1 - 6 - 7 471 59
AF289584.1 - 6 - 7 742 59
AF358135.1 - 6 - 7 473 59
AK314930.1 - 6 - 7 624 59
AY061883.1 - 6 - 7 624 59
BC051288.1 - 6 - 7 690 59

View Full Product Details