The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001798.4 | NP_001789.2 | 2 - 3 | 432 | 148 |
NM_052827.3 | NP_439892.2 | 2 - 3 | 432 | 148 | |
XM_011537732.1 | XP_011536034.1 | 2 - 3 | 432 | 148 | |
GenBank mRNA | AB012305.1 | - | 2 - 3 | 194 | 148 |
AK291941.1 | - | 2 - 3 | 371 | 148 | |
BC003065.2 | - | 2 - 3 | 352 | 148 | |
BJ991087.1 | - | 2 - 3 | 429 | 148 | |
BT006821.1 | - | 2 - 3 | 192 | 148 | |
DA814453.1 | - | 2 - 3 | 432 | 148 | |
M68520.1 | - | 2 - 3 | 382 | 148 | |
X61622.1 | - | 2 - 3 | 192 | 148 | |
X62071.1 | - | 2 - 3 | 362 | 148 |