Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPFIA1
Entrez Gene ID
8500
Gene Name
PTPRF interacting protein alpha 1
Gene Aliases
LIP.1, LIP1, LIPRIN
Chromosome Location
Chr.11: 70270687 - 70384501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003626.3 NP_003617.1 10 - 11 1507 103
NM_177423.2 NP_803172.1 10 - 11 1507 103
NR_045286.1 10 - 11 1507 103
XM_006718716.3 XP_006718779.1 11 - 12 1595 103
XM_011545306.2 XP_011543608.1 11 - 12 1467 103
XM_011545307.2 XP_011543609.1 11 - 12 1595 103
XM_011545308.2 XP_011543610.1 11 - 12 1595 103
XM_011545309.2 XP_011543611.1 11 - 12 1595 103
XM_011545310.2 XP_011543612.1 10 - 11 1520 103
XM_011545311.2 XP_011543613.1 11 - 12 1595 103
XM_011545312.2 XP_011543614.1 11 - 12 1595 103
XM_011545313.2 XP_011543615.1 11 - 12 1595 103
XM_011545314.2 XP_011543616.1 11 - 12 1595 103
XM_011545315.2 XP_011543617.1 10 - 11 1520 103
XM_011545316.2 XP_011543618.1 11 - 12 1595 103
XM_011545317.2 XP_011543619.1 11 - 12 1595 103
XM_011545319.1 XP_011543621.1 10 - 11 1520 103
XM_017018448.1 XP_016873937.1 11 - 12 1595 103
XM_017018449.1 XP_016873938.1 10 - 11 1520 103
XM_017018450.1 XP_016873939.1 11 - 12 1595 103
GenBank mRNA AB209314.1 - 10 - 11 1434 103
BC034046.1 - 10 - 11 1470 103
BX648037.1 - 6 - 7 679 103
U22815.1 - 10 - 11 1521 103
U22816.1 - 10 - 11 1521 103

View Full Product Details