Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPFIA1
Entrez Gene ID
8500
Gene Name
PTPRF interacting protein alpha 1
Gene Aliases
LIP.1, LIP1, LIPRIN
Chromosome Location
Chr.11: 70270687 - 70384501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003626.3 NP_003617.1 15 - 16 2150 66
NM_177423.2 NP_803172.1 15 - 16 2150 66
NR_045286.1 15 - 16 2131 66
XM_006718716.3 XP_006718779.1 16 - 17 2238 66
XM_011545306.2 XP_011543608.1 16 - 17 2110 66
XM_011545307.2 XP_011543609.1 16 - 17 2238 66
XM_011545308.2 XP_011543610.1 16 - 17 2238 66
XM_011545309.2 XP_011543611.1 16 - 17 2238 66
XM_011545310.2 XP_011543612.1 15 - 16 2163 66
XM_011545311.2 XP_011543613.1 16 - 17 2238 66
XM_011545312.2 XP_011543614.1 16 - 17 2238 66
XM_011545313.2 XP_011543615.1 16 - 17 2238 66
XM_011545314.2 XP_011543616.1 16 - 17 2238 66
XM_011545315.2 XP_011543617.1 15 - 16 2163 66
XM_011545316.2 XP_011543618.1 16 - 17 2238 66
XM_011545317.2 XP_011543619.1 16 - 17 2238 66
XM_011545318.2 XP_011543620.1 2 - 3 460 66
XM_011545319.1 XP_011543621.1 15 - 16 2163 66
XM_017018448.1 XP_016873937.1 16 - 17 2238 66
XM_017018449.1 XP_016873938.1 15 - 16 2163 66
XM_017018450.1 XP_016873939.1 16 - 17 2238 66
GenBank mRNA AB209314.1 - 15 - 16 2058 66
BC034046.1 - 15 - 16 2113 66
BX648037.1 - 11 - 12 1327 66
U22815.1 - 15 - 16 2164 66
U22816.1 - 15 - 16 2164 66

View Full Product Details