The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198700.2 | NP_941989.1 | 1 - 2 | 197 | 110 |
XM_011519854.1 | XP_011518156.1 | 5 - 6 | 534 | 110 | |
XM_011519859.1 | XP_011518161.1 | 5 - 6 | 534 | 110 | |
XM_017017124.1 | XP_016872613.1 | 3 - 4 | 349 | 110 | |
GenBank mRNA | BC031079.1 | - | 1 - 2 | 197 | 110 |