The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318339.1 | NP_001305268.1 | 10 - 11 | 1884 | 101 |
NM_015308.3 | NP_056123.2 | 10 - 11 | 1878 | 101 | |
GenBank mRNA | AB023231.2 | - | 10 - 11 | 1877 | 101 |
AK022987.1 | - | 3 - 4 | 322 | 101 | |
AK054636.1 | - | 10 - 11 | 1701 | 101 | |
AK123898.1 | - | 9 - 10 | 1539 | 101 | |
AL137480.1 | - | 10 - 11 | 1677 | 101 | |
BC037404.1 | - | 10 - 11 | 1709 | 101 |