Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNBP4
Entrez Gene ID
23360
Gene Name
formin binding protein 4
Gene Aliases
FBP30
Chromosome Location
Chr.11: 47716510 - 47767478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318339.1 NP_001305268.1 11 - 12 2013 111
NM_015308.3 NP_056123.2 11 - 12 2007 111
GenBank mRNA AB023231.2 - 11 - 12 2006 111
AK022987.1 - 4 - 5 451 111
AK054636.1 - 11 - 12 1830 111
AL137480.1 - 11 - 12 1806 111
BC037404.1 - 11 - 12 1838 111

View Full Product Details