Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNBP4
Entrez Gene ID
23360
Gene Name
formin binding protein 4
Gene Aliases
FBP30
Chromosome Location
Chr.11: 47716510 - 47767478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318339.1 NP_001305268.1 9 - 10 1778 122
NM_015308.3 NP_056123.2 9 - 10 1772 122
GenBank mRNA AB023231.2 - 9 - 10 1771 122
AK022987.1 - 2 - 3 216 122
AK054636.1 - 9 - 10 1595 122
AL137480.1 - 9 - 10 1571 122
BC037404.1 - 9 - 10 1603 122

View Full Product Details