The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025130.3 | NP_079406.3 | 11 - 12 | 1848 | 94 |
XM_011540195.2 | XP_011538497.1 | 11 - 12 | 1848 | 94 | |
GenBank mRNA | AK026414.1 | - | 6 - 7 | 1128 | 94 |
AK095086.1 | - | 11 - 12 | 1848 | 94 | |
AK291024.1 | - | 11 - 12 | 1815 | 94 | |
AK303408.1 | - | 8 - 9 | 1351 | 94 | |
BC012337.1 | - | 5 - 6 | 997 | 94 | |
BC021278.2 | - | 3 - 4 | 554 | 94 | |
BC110504.1 | - | 11 - 12 | 1844 | 94 | |
BC110505.2 | - | 11 - 12 | 1847 | 94 |