The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025130.3 | NP_079406.3 | 13 - 14 | 2070 | 71 |
XM_011540195.2 | XP_011538497.1 | 13 - 14 | 2070 | 71 | |
GenBank mRNA | AK026414.1 | - | 8 - 9 | 1350 | 71 |
AK095086.1 | - | 13 - 14 | 2070 | 71 | |
AK291024.1 | - | 13 - 14 | 2037 | 71 | |
AK303408.1 | - | 10 - 11 | 1573 | 71 | |
BC012337.1 | - | 7 - 8 | 1219 | 71 | |
BC021278.2 | - | 5 - 6 | 776 | 71 | |
BC110504.1 | - | 13 - 14 | 2066 | 71 | |
BC110505.2 | - | 13 - 14 | 2069 | 71 |