The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276435.1 | NP_001263364.1 | 2 - 3 | 226 | 84 |
NM_001276437.1 | NP_001263366.1 | 2 - 3 | 187 | 84 | |
NM_001276439.1 | NP_001263368.1 | 2 - 3 | 245 | 84 | |
NM_002243.4 | NP_002234.2 | 2 - 3 | 226 | 84 | |
NM_170736.2 | NP_733932.1 | 1 - 2 | 378 | 84 | |
GenBank mRNA | AK292812.1 | - | 1 - 2 | 113 | 84 |
BC013327.1 | - | 2 - 3 | 186 | 84 | |
D87291.1 | - | 1 - 2 | 234 | 84 | |
DA670230.1 | - | 2 - 3 | 299 | 84 | |
DA673158.1 | - | 2 - 3 | 245 | 84 | |
DA968871.1 | - | 2 - 3 | 226 | 84 |