Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA5A
Entrez Gene ID
9037
Gene Name
semaphorin 5A
Gene Aliases
SEMAF, semF
Chromosome Location
Chr.5: 9035026 - 9546121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003966.2 NP_003957.2 18 - 19 104
XM_006714506.2 XP_006714569.1 19 - 20 104
XM_006714507.3 XP_006714570.1 17 - 18 104
XM_011514155.2 XP_011512457.1 18 - 19 104
XM_011514156.1 XP_011512458.1 20 - 21 104
XM_011514157.1 XP_011512459.1 19 - 20 104
XM_011514158.1 XP_011512460.1 17 - 18 104
XM_011514159.2 XP_011512461.1 14 - 15 104
XM_017010016.1 XP_016865505.1 17 - 18 104
GenBank mRNA AK293441.1 - 8 - 9 104
AK307587.1 - 18 - 19 104
BC115696.1 - 18 - 19 104
KJ534937.1 - 16 - 17 104
KJ535077.1 - 15 - 16 104
U52840.1 - 18 - 19 3152 104

View Full Product Details