The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003802.2 | NP_003793.2 | 27 - 28 | 3825 | 65 |
GenBank mRNA | AF111782.2 | - | 27 - 28 | 3825 | 65 |