The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003802.2 | NP_003793.2 | 33 - 34 | 4750 | 61 |
GenBank mRNA | AF111782.2 | - | 33 - 34 | 4750 | 61 |