The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167674.1 | NP_001161146.1 | 4 - 5 | 1119 | 84 |
NM_001167675.1 | NP_001161147.1 | 5 - 6 | 1146 | 84 | |
NM_001256502.1 | NP_001243431.1 | 5 - 6 | 666 | 84 | |
NM_001256503.1 | NP_001243432.1 | 4 - 5 | 516 | 84 | |
NM_001256504.1 | NP_001243433.1 | 6 - 7 | 693 | 84 | |
NM_001256505.1 | NP_001243434.1 | 5 - 6 | 666 | 84 | |
NM_153184.3 | NP_694854.2 | 4 - 5 | 499 | 84 | |
XM_006713081.3 | XP_006713144.1 | 5 - 6 | 1009 | 84 | |
XM_006713082.3 | XP_006713145.1 | 4 - 5 | 1112 | 84 | |
XM_017006062.1 | XP_016861551.1 | 6 - 7 | 1051 | 84 | |
XM_017006063.1 | XP_016861552.1 | 4 - 5 | 699 | 84 | |
GenBank mRNA | AB646739.1 | - | 5 - 6 | 1146 | 84 |
AB646740.1 | - | 5 - 6 | 666 | 84 | |
AB646741.1 | - | 5 - 6 | 666 | 84 | |
AB646742.1 | - | 6 - 7 | 693 | 84 | |
AB646743.1 | - | 4 - 5 | 516 | 84 | |
AK295885.1 | - | 5 - 6 | 1142 | 84 | |
AL833049.1 | - | 2 - 3 | 183 | 84 | |
AL834270.1 | - | 4 - 5 | 1089 | 84 | |
BC105999.1 | - | 5 - 6 | 1146 | 84 | |
BC112136.1 | - | 4 - 5 | 555 | 84 | |
BC112138.1 | - | 4 - 5 | 567 | 84 |