Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELL
Entrez Gene ID
8178
Gene Name
elongation factor for RNA polymerase II
Gene Aliases
C19orf17, ELL1, MEN, PPP1R68
Chromosome Location
Chr.19: 18442663 - 18522129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006532.3 NP_006523.1 5 - 6 814 60
XM_011528330.2 XP_011526632.2 5 - 6 816 60
XM_017027335.1 XP_016882824.1 6 - 7 798 60
GenBank mRNA AB208794.1 - 6 - 7 795 60
AK225481.1 - 5 - 6 753 60
AK293105.1 - 5 - 6 747 60
BC049195.1 - 5 - 6 796 60
U16282.1 - 5 - 6 754 60

View Full Product Details