The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006532.3 | NP_006523.1 | 9 - 10 | 1610 | 59 |
XM_017027335.1 | XP_016882824.1 | 10 - 11 | 1594 | 59 | |
XM_017027336.1 | XP_016882825.1 | 5 - 6 | 1346 | 59 | |
XM_017027337.1 | XP_016882826.1 | 4 - 5 | 786 | 59 | |
GenBank mRNA | AB208794.1 | - | 10 - 11 | 1591 | 59 |
AK293105.1 | - | 9 - 10 | 1543 | 59 | |
BC049195.1 | - | 9 - 10 | 1592 | 59 | |
U16282.1 | - | 9 - 10 | 1550 | 59 |