The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014062.2 | NP_054781.1 | 1 - 2 | 120 | 79 |
NR_074074.1 | 1 - 2 | 120 | 79 | ||
GenBank mRNA | AB026125.1 | - | 1 - 2 | 78 | 79 |
AF190161.1 | - | 1 - 2 | 48 | 79 | |
AK001028.1 | - | 1 - 2 | 76 | 79 | |
AK314307.1 | - | 1 - 2 | 75 | 79 | |
AY487344.1 | - | 1 - 2 | 94 | 79 | |
BC064630.1 | - | 1 - 2 | 63 | 79 | |
BG386829.1 | - | 1 - 1 | 64 | 79 | |
BM827373.1 | - | 1 - 2 | 96 | 79 | |
DA004856.1 | - | 1 - 2 | 76 | 79 | |
DB048531.1 | - | 1 - 2 | 120 | 79 |