Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CES4A
- Entrez Gene ID
-
283848
- Gene Name
- carboxylesterase 4A
- Gene Aliases
-
CES6, CES8
- Chromosome Location
-
Chr.16: 66988589 - 67009758 on Build GRCh38
View Full Product Details