The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152319.3 | NP_689532.1 | 6 - 7 | 323 | 67 |
XM_005268636.3 | XP_005268693.1 | 5 - 6 | 205 | 67 | |
XM_011537896.2 | XP_011536198.1 | 8 - 9 | 971 | 67 | |
XM_017018796.1 | XP_016874285.1 | 10 - 11 | 1161 | 67 | |
GenBank mRNA | AY255793.1 | - | 5 - 6 | 228 | 67 |
BC031670.2 | - | 5 - 6 | 249 | 67 | |
BQ429029.1 | - | 6 - 7 | 326 | 67 |