The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024322.3 | NP_077298.1 | 1 - 2 | 410 | 64 |
GenBank mRNA | AK315483.1 | - | 1 - 2 | 36 | 64 |
BC002870.1 | - | 1 - 2 | 67 | 64 | |
BP366489.1 | - | 1 - 2 | 63 | 64 |