Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1C
Entrez Gene ID
775
Gene Name
calcium voltage-gated channel subunit alpha1 C
Gene Aliases
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location
Chr.12: 1969677 - 2697949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000719.6 NP_000710.5 1 - 2 365 106
NM_001129827.1 NP_001123299.1 1 - 2 365 106
NM_001129829.1 NP_001123301.1 1 - 2 365 106
NM_001129830.2 NP_001123302.2 1 - 2 330 106
NM_001129831.1 NP_001123303.1 1 - 2 365 106
NM_001129832.1 NP_001123304.1 1 - 2 365 106
NM_001129833.1 NP_001123305.1 1 - 2 365 106
NM_001129834.1 NP_001123306.1 1 - 2 365 106
NM_001129835.1 NP_001123307.1 1 - 2 365 106
NM_001129836.1 NP_001123308.1 1 - 2 365 106
NM_001129837.1 NP_001123309.1 1 - 2 365 106
NM_001129838.1 NP_001123310.1 1 - 2 365 106
NM_001129839.1 NP_001123311.1 1 - 2 365 106
NM_001129840.1 NP_001123312.1 1 - 2 365 106
NM_001129841.1 NP_001123313.1 1 - 2 365 106
NM_001129842.1 NP_001123314.1 1 - 2 365 106
NM_001129843.1 NP_001123315.1 1 - 2 365 106
NM_001129844.1 NP_001123316.1 1 - 2 365 106
NM_001129846.1 NP_001123318.1 1 - 2 365 106
NM_001167623.1 NP_001161095.1 1 - 2 365 106
NM_001167624.2 NP_001161096.2 1 - 2 330 106
NM_001167625.1 NP_001161097.1 1 - 2 365 106
NM_199460.3 NP_955630.3 1 - 2 330 106
XM_011521023.2 XP_011519325.1 1 - 2 604 106
GenBank mRNA AB209016.1 - 1 - 2 77 106
AF465484.1 - 1 - 2 317 106
AK308652.1 - 1 - 2 289 106
AK309111.1 - 1 - 2 330 106
BC146846.1 - 1 - 2 180 106

View Full Product Details