The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015512.4 | NP_056327.4 | 52 - 53 | 8585 | 106 |
XM_017006129.1 | XP_016861618.1 | 54 - 55 | 11542 | 106 | |
XM_017006130.1 | XP_016861619.1 | 53 - 54 | 11472 | 106 | |
XM_017006131.1 | XP_016861620.1 | 54 - 55 | 11542 | 106 | |
XM_017006132.1 | XP_016861621.1 | 54 - 55 | 11543 | 106 | |
XM_017006133.1 | XP_016861622.1 | 54 - 55 | 11543 | 106 | |
GenBank mRNA | AB037831.3 | - | 52 - 53 | 8585 | 106 |
AB290163.1 | - | 52 - 53 | 8585 | 106 | |
AY221994.1 | - | 9 - 10 | 1433 | 106 |