The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015512.4 | NP_056327.4 | 72 - 73 | 11870 | 68 |
XM_017006129.1 | XP_016861618.1 | 74 - 75 | 14827 | 68 | |
XM_017006130.1 | XP_016861619.1 | 73 - 74 | 14757 | 68 | |
XM_017006131.1 | XP_016861620.1 | 73 - 74 | 14701 | 68 | |
GenBank mRNA | AB037831.3 | - | 72 - 73 | 11870 | 68 |
AB290163.1 | - | 71 - 72 | 12065 | 68 | |
AK023766.1 | - | 12 - 13 | 1880 | 68 | |
AK074130.2 | - | 19 - 20 | 3395 | 68 | |
AL117428.1 | - | 4 - 5 | 589 | 68 | |
AY221994.1 | - | 28 - 29 | 4913 | 68 |