Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CARF
- Entrez Gene ID
-
79800
- Gene Name
- calcium responsive transcription factor
- Gene Aliases
-
ALS2CR8, NYD-SP24
- Chromosome Location
-
Chr.2: 202912218 - 202987856 on Build GRCh38
View Full Product Details