The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104586.2 | NP_001098056.1 | 3 - 4 | 397 | 69 |
NM_001322427.1 | NP_001309356.1 | 4 - 5 | 456 | 69 | |
NM_001322428.1 | NP_001309357.1 | 3 - 4 | 670 | 69 | |
NM_001322430.1 | NP_001309359.1 | 3 - 4 | 397 | 69 | |
NM_024744.15 | NP_079020.13 | 4 - 5 | 1294 | 69 | |
NR_136330.1 | 3 - 4 | 397 | 69 | ||
NR_136331.1 | 3 - 4 | 397 | 69 | ||
XM_005246858.3 | XP_005246915.1 | 4 - 5 | 403 | 69 | |
XM_005246859.3 | XP_005246916.1 | 4 - 5 | 455 | 69 | |
XM_006712760.2 | XP_006712823.1 | 3 - 4 | 338 | 69 | |
XM_011511867.2 | XP_011510169.1 | 4 - 5 | 434 | 69 | |
XM_011511868.2 | XP_011510170.1 | 3 - 4 | 338 | 69 | |
XM_017004961.1 | XP_016860450.1 | 3 - 4 | 792 | 69 | |
GenBank mRNA | AB053309.1 | - | 4 - 5 | 777 | 69 |
AF454946.1 | - | 1 - 2 | 124 | 69 | |
AK299366.1 | - | 3 - 4 | 332 | 69 | |
AK308171.1 | - | 3 - 4 | 337 | 69 | |
AL834188.1 | - | 3 - 4 | 360 | 69 | |
AY032876.2 | - | 3 - 4 | 349 | 69 | |
BC033777.2 | - | 3 - 4 | 410 | 69 |