The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271999.1 | NP_001258928.1 | 12 - 13 | 1531 | 130 |
NM_001272000.1 | NP_001258929.1 | 12 - 13 | 1531 | 130 | |
NM_001272001.1 | NP_001258930.1 | 12 - 13 | 1531 | 130 | |
NM_001272002.1 | NP_001258931.1 | 12 - 13 | 1531 | 130 | |
NM_033407.3 | NP_212132.2 | 12 - 13 | 1531 | 130 | |
XM_005271292.2 | XP_005271349.1 | 12 - 13 | 1521 | 130 | |
XM_011542326.2 | XP_011540628.1 | 12 - 13 | 1521 | 130 | |
XM_011542327.2 | XP_011540629.1 | 12 - 13 | 1521 | 130 | |
XM_011542328.2 | XP_011540630.1 | 12 - 13 | 1521 | 130 | |
XM_011542330.2 | XP_011540632.1 | 12 - 13 | 1521 | 130 | |
XM_017002639.1 | XP_016858128.1 | 12 - 13 | 1521 | 130 | |
XM_017002640.1 | XP_016858129.1 | 12 - 13 | 1521 | 130 | |
GenBank mRNA | AK055401.1 | - | 12 - 13 | 1461 | 130 |
AK292640.1 | - | 12 - 13 | 1521 | 130 | |
DQ118679.1 | - | 9 - 10 | 1103 | 130 | |
DQ118680.1 | - | 9 - 10 | 1103 | 130 | |
DQ309763.1 | - | 9 - 10 | 1103 | 130 | |
DQ341187.1 | - | 12 - 13 | 1438 | 130 |