Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMBX1
Entrez Gene ID
127343
Gene Name
diencephalon/mesencephalon homeobox 1
Gene Aliases
MBX, OTX3, PAXB
Chromosome Location
Chr.1: 46489837 - 46514226 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_147192.2 NP_671725.1 2 - 3 363 89
NM_172225.1 NP_757379.1 2 - 3 348 89
XM_011540668.2 XP_011538970.1 3 - 4 594 89
XM_017000289.1 XP_016855778.1 3 - 4 386 89
GenBank mRNA AB037699.1 - 2 - 3 348 89
AF398527.1 - 2 - 3 339 89
AF398528.1 - 2 - 3 354 89
AY726596.1 - 1 - 2 173 89

View Full Product Details