The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015071.4 | NP_055886.1 | 21 - 22 | 2302 | 67 |
XM_005268398.4 | XP_005268455.1 | 21 - 22 | 2539 | 67 | |
XM_017009247.1 | XP_016864736.1 | 22 - 23 | 2616 | 67 | |
XM_017009248.1 | XP_016864737.1 | 22 - 23 | 2403 | 67 | |
GenBank mRNA | AB014521.1 | - | 20 - 21 | 2084 | 67 |
KU178562.1 | - | 21 - 22 | 2082 | 67 |