The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293557.1 | NP_001280486.1 | 1 - 2 | 606 | 68 |
NM_022162.2 | NP_071445.1 | 2 - 3 | 646 | 68 | |
XM_005256084.3 | XP_005256141.1 | 2 - 3 | 550 | 68 | |
XM_006721242.3 | XP_006721305.1 | 2 - 3 | 550 | 68 | |
XM_006721243.3 | XP_006721306.1 | 2 - 3 | 550 | 68 | |
XM_011523260.2 | XP_011521562.1 | 2 - 3 | 550 | 68 | |
XM_011523261.1 | XP_011521563.1 | 2 - 3 | 550 | 68 | |
GenBank mRNA | AF178930.1 | - | 2 - 3 | 646 | 68 |
AY187243.1 | - | 2 - 3 | 646 | 68 | |
AY187244.1 | - | 2 - 3 | 68 | ||
DQ868973.1 | - | 2 - 3 | 528 | 68 | |
HQ204571.1 | - | 2 - 3 | 528 | 68 |