The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002463.1 | NP_002454.1 | 10 - 11 | 1513 | 92 |
XM_005260983.4 | XP_005261040.1 | 10 - 11 | 1611 | 92 | |
XM_005260984.1 | XP_005261041.1 | 10 - 11 | 1791 | 92 | |
XM_011529571.1 | XP_011527873.1 | 10 - 11 | 1546 | 92 | |
XM_011529572.1 | XP_011527874.1 | 10 - 11 | 1649 | 92 | |
XM_011529573.1 | XP_011527875.1 | 9 - 10 | 1415 | 92 | |
XM_011529574.2 | XP_011527876.1 | 5 - 6 | 693 | 92 | |
GenBank mRNA | AK122952.1 | - | 5 - 6 | 1699 | 92 |
BC035293.1 | - | 9 - 10 | 1475 | 92 | |
M30818.1 | - | 10 - 11 | 1513 | 92 | |
M33883.1 | - | 9 - 10 | 1326 | 92 |