Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MX2
Entrez Gene ID
4600
Gene Name
MX dynamin like GTPase 2
Gene Aliases
MXB
Chromosome Location
Chr.21: 41361986 - 41408943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002463.1 NP_002454.1 13 - 14 2004 63
XM_005260983.4 XP_005261040.1 13 - 14 2102 63
XM_005260984.1 XP_005261041.1 13 - 14 2282 63
XM_011529571.1 XP_011527873.1 13 - 14 2037 63
XM_011529572.1 XP_011527874.1 13 - 14 2140 63
XM_011529573.1 XP_011527875.1 12 - 13 1906 63
XM_011529574.2 XP_011527876.1 8 - 9 1184 63
GenBank mRNA AK122952.1 - 8 - 9 2190 63
AK127895.1 - 2 - 3 1253 63
AY312582.1 - 4 - 5 521 63
BC035293.1 - 12 - 13 1966 63
M30818.1 - 13 - 14 2004 63
M33883.1 - 12 - 13 1817 63

View Full Product Details