The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002463.1 | NP_002454.1 | 7 - 8 | 1172 | 124 |
XM_005260983.4 | XP_005261040.1 | 7 - 8 | 1270 | 124 | |
XM_005260984.1 | XP_005261041.1 | 7 - 8 | 1450 | 124 | |
XM_011529571.1 | XP_011527873.1 | 7 - 8 | 1205 | 124 | |
XM_011529572.1 | XP_011527874.1 | 7 - 8 | 1308 | 124 | |
XM_011529573.1 | XP_011527875.1 | 6 - 7 | 1074 | 124 | |
XM_011529574.2 | XP_011527876.1 | 2 - 3 | 352 | 124 | |
GenBank mRNA | AK122952.1 | - | 2 - 3 | 1358 | 124 |
AK298780.1 | - | 5 - 6 | 1130 | 124 | |
BC035293.1 | - | 6 - 7 | 1134 | 124 | |
M30818.1 | - | 7 - 8 | 1172 | 124 | |
M33883.1 | - | 6 - 7 | 985 | 124 |