The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022365.3 | NP_071760.2 | 2 - 3 | 612 | 138 |
XM_011519614.2 | XP_011517916.1 | 2 - 3 | 659 | 138 | |
XM_017016536.1 | XP_016872025.1 | 2 - 3 | 659 | 138 | |
GenBank mRNA | AK027263.1 | - | 2 - 3 | 432 | 138 |
AK308369.1 | - | 2 - 3 | 618 | 138 | |
AY225122.1 | - | 2 - 3 | 325 | 138 | |
BC110894.1 | - | 2 - 3 | 612 | 138 |