Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC1
Entrez Gene ID
64215
Gene Name
DnaJ heat shock protein family (Hsp40) member C1
Gene Aliases
DNAJL1, ERdj1, HTJ1, MTJ1
Chromosome Location
Chr.10: 21756548 - 22003768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022365.3 NP_071760.2 6 - 7 1011 112
XM_011519614.2 XP_011517916.1 6 - 7 1058 112
XM_017016536.1 XP_016872025.1 6 - 7 1058 112
GenBank mRNA AK026062.1 - 3 - 4 284 112
AK027263.1 - 6 - 7 831 112
AK308369.1 - 6 - 7 1017 112
AY225122.1 - 6 - 7 724 112
BC030955.1 - 2 - 3 194 112
BC110894.1 - 6 - 7 1011 112

View Full Product Details