The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100396.1 | NP_001093866.1 | 2 - 3 | 279 | 151 |
XM_011532510.2 | XP_011530812.1 | 2 - 3 | 349 | 151 | |
XM_017003324.1 | XP_016858813.1 | 2 - 3 | 353 | 151 | |
GenBank mRNA | BC031669.1 | - | 2 - 3 | 242 | 151 |