Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF18
Entrez Gene ID
116441
Gene Name
transmembrane 4 L six family member 18
Gene Aliases
L6D
Chromosome Location
Chr.3: 149318498 - 149333761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184723.1 NP_001171652.1 1 - 2 265 137
NM_138786.3 NP_620141.1 2 - 3 441 137
GenBank mRNA AK313400.1 - 2 - 3 215 137
AL833134.1 - 2 - 3 364 137
BC014339.1 - 2 - 3 317 137
BF213446.1 - - 276 137
CA454331.1 - 1 - 2 227 137

View Full Product Details