The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002373.5 | NP_002364.5 | 5 - 6 | 8726 | 93 |
XM_017022189.1 | XP_016877678.1 | 6 - 7 | 9016 | 93 | |
GenBank mRNA | AI124707.1 | - | 2 - 3 | 313 | 93 |