Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
Chromosome Location
Chr.20: 62952647 - 62968597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302643.1 NP_001289572.1 11 - 12 1311 56
NM_022082.3 NP_071365.3 10 - 11 1249 56
XM_011528978.2 XP_011527280.1 10 - 11 1161 56
GenBank mRNA AK027065.1 - 3 - 4 238 56
AK074107.1 - 9 - 10 2045 56
AK095473.1 - 10 - 11 1100 56
AK125597.1 - 10 - 11 1772 56
AK298503.1 - 10 - 11 1155 56
BC025312.1 - 11 - 12 1251 56
BC027447.1 - 6 - 7 470 56
BC038593.1 - 10 - 11 1091 56

View Full Product Details