The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302643.1 | NP_001289572.1 | 3 - 4 | 504 | 76 |
NM_022082.3 | NP_071365.3 | 2 - 3 | 442 | 76 | |
GenBank mRNA | AK074107.1 | - | 2 - 3 | 378 | 76 |
AK095473.1 | - | 2 - 3 | 293 | 76 | |
AK125597.1 | - | 3 - 4 | 447 | 76 | |
AK298503.1 | - | 2 - 3 | 348 | 76 | |
BC025312.1 | - | 3 - 4 | 444 | 76 | |
BC038593.1 | - | 2 - 3 | 284 | 76 | |
DB153870.1 | - | 2 - 3 | 442 | 76 |