The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289934.1 | NP_001276863.1 | 11 - 12 | 1386 | 125 |
NM_002319.4 | NP_002310.2 | 11 - 12 | 1386 | 125 | |
GenBank mRNA | AK074833.1 | - | 11 - 12 | 1341 | 125 |
AK097420.1 | - | 11 - 12 | 1216 | 125 | |
AK302410.1 | - | 11 - 12 | 1281 | 125 | |
BC007634.1 | - | 10 - 11 | 990 | 125 | |
BC018529.1 | - | 11 - 12 | 1351 | 125 |