Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRD
Entrez Gene ID
5789
Gene Name
protein tyrosine phosphatase, receptor type D
Gene Aliases
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location
Chr.9: 8314246 - 10612840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040712.2 NP_001035802.1 19 - 20 2961 112
NM_001171025.1 NP_001164496.1 18 - 19 2970 112
NM_002839.3 NP_002830.1 35 - 36 4831 112
NM_130391.3 NP_569075.2 20 - 21 2970 112
NM_130392.3 NP_569076.2 20 - 21 2973 112
NM_130393.3 NP_569077.2 18 - 19 2943 112
XM_006716817.3 XP_006716880.1 36 - 37 5180 112
XM_006716823.2 XP_006716886.1 34 - 35 5126 112
XM_006716825.3 XP_006716888.1 34 - 35 5125 112
XM_006716827.3 XP_006716890.1 31 - 32 5065 112
XM_006716832.3 XP_006716895.1 28 - 29 3896 112
XM_006716833.3 XP_006716896.1 30 - 31 3898 112
XM_006716834.3 XP_006716897.1 28 - 29 3899 112
XM_006716835.3 XP_006716898.1 29 - 30 3889 112
XM_006716837.3 XP_006716900.1 28 - 29 3869 112
XM_006716838.3 XP_006716901.1 28 - 29 3868 112
XM_006716839.3 XP_006716902.1 27 - 28 3854 112
XM_011517992.2 XP_011516294.1 34 - 35 5122 112
XM_017014958.1 XP_016870447.1 36 - 37 5188 112
XM_017014959.1 XP_016870448.1 37 - 38 5260 112
XM_017014960.1 XP_016870449.1 35 - 36 5149 112
XM_017014961.1 XP_016870450.1 35 - 36 5133 112
XM_017014962.1 XP_016870451.1 27 - 28 4321 112
XM_017014963.1 XP_016870452.1 36 - 37 5185 112
XM_017014964.1 XP_016870453.1 36 - 37 5184 112
XM_017014965.1 XP_016870454.1 36 - 37 5187 112
XM_017014966.1 XP_016870455.1 35 - 36 5179 112
XM_017014967.1 XP_016870456.1 35 - 36 5174 112
XM_017014968.1 XP_016870457.1 35 - 36 5170 112
XM_017014969.1 XP_016870458.1 35 - 36 5170 112
XM_017014970.1 XP_016870459.1 35 - 36 5170 112
XM_017014971.1 XP_016870460.1 34 - 35 5159 112
XM_017014972.1 XP_016870461.1 35 - 36 5161 112
XM_017014973.1 XP_016870462.1 34 - 35 5155 112
XM_017014974.1 XP_016870463.1 35 - 36 5158 112
XM_017014975.1 XP_016870464.1 33 - 34 5144 112
XM_017014976.1 XP_016870465.1 35 - 36 5143 112
XM_017014977.1 XP_016870466.1 34 - 35 5144 112
XM_017014978.1 XP_016870467.1 35 - 36 5140 112
XM_017014979.1 XP_016870468.1 35 - 36 5139 112
XM_017014980.1 XP_016870469.1 33 - 34 5095 112
XM_017014981.1 XP_016870470.1 30 - 31 5051 112
XM_017014982.1 XP_016870471.1 31 - 32 3951 112
XM_017014983.1 XP_016870472.1 31 - 32 3948 112
XM_017014984.1 XP_016870473.1 30 - 31 3939 112
XM_017014985.1 XP_016870474.1 30 - 31 3933 112
XM_017014986.1 XP_016870475.1 30 - 31 3929 112
XM_017014987.1 XP_016870476.1 29 - 30 3914 112
XM_017014988.1 XP_016870477.1 29 - 30 3911 112
XM_017014989.1 XP_016870478.1 30 - 31 3905 112
XM_017014990.1 XP_016870479.1 30 - 31 3901 112
XM_017014991.1 XP_016870480.1 30 - 31 3901 112
XM_017014992.1 XP_016870481.1 29 - 30 3892 112
XM_017014993.1 XP_016870482.1 29 - 30 3887 112
XM_017014994.1 XP_016870483.1 29 - 30 3884 112
XM_017014995.1 XP_016870484.1 29 - 30 3890 112
GenBank mRNA AB208869.1 - 18 - 19 2787 112
AB211400.1 - 34 - 35 4799 112
AK296392.1 - 3 - 4 228 112
BC106713.1 - 18 - 19 2918 112
BC106714.1 - 20 - 21 2921 112
BC106715.2 - 20 - 21 2918 112
BC106716.1 - 18 - 19 2891 112
L38929.1 - 25 - 26 4241 112
X54133.1 - 15 - 16 2921 112

View Full Product Details