Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG2
Entrez Gene ID
1740
Gene Name
discs large MAGUK scaffold protein 2
Gene Aliases
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location
Chr.11: 83455009 - 85628534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142699.1 NP_001136171.1 18 - 19 2139 76
NM_001142700.1 NP_001136172.1 10 - 11 1228 76
NM_001206769.1 NP_001193698.1 12 - 13 1823 76
NM_001300983.1 NP_001287912.1 13 - 14 1815 76
NM_001364.3 NP_001355.2 13 - 14 1815 76
XM_005273810.4 XP_005273867.1 12 - 13 1458 76
XM_005273811.4 XP_005273868.1 12 - 13 1458 76
XM_011544778.2 XP_011543080.1 19 - 20 3430 76
XM_011544780.2 XP_011543082.1 19 - 20 3430 76
XM_011544782.2 XP_011543084.1 14 - 15 1901 76
XM_011544788.2 XP_011543090.1 11 - 12 1302 76
XM_017017254.1 XP_016872743.1 19 - 20 3430 76
XM_017017255.1 XP_016872744.1 18 - 19 3391 76
XM_017017256.1 XP_016872745.1 18 - 19 2920 76
XM_017017257.1 XP_016872746.1 17 - 18 3852 76
XM_017017258.1 XP_016872747.1 16 - 17 3028 76
XM_017017259.1 XP_016872748.1 17 - 18 2845 76
XM_017017260.1 XP_016872749.1 16 - 17 3777 76
XM_017017261.1 XP_016872750.1 18 - 19 3274 76
XM_017017262.1 XP_016872751.1 12 - 13 1883 76
XM_017017263.1 XP_016872752.1 14 - 15 1901 76
XM_017017264.1 XP_016872753.1 13 - 14 2078 76
XM_017017265.1 XP_016872754.1 13 - 14 2075 76
XM_017017266.1 XP_016872755.1 12 - 13 1881 76
XM_017017267.1 XP_016872756.1 13 - 14 1983 76
XM_017017268.1 XP_016872757.1 13 - 14 1804 76
XM_017017269.1 XP_016872758.1 13 - 14 2505 76
XM_017017270.1 XP_016872759.1 14 - 15 1901 76
XM_017017271.1 XP_016872760.1 13 - 14 2078 76
XM_017017272.1 XP_016872761.1 13 - 14 2075 76
XM_017017273.1 XP_016872762.1 11 - 12 1734 76
XM_017017274.1 XP_016872763.1 13 - 14 1983 76
XM_017017275.1 XP_016872764.1 11 - 12 1726 76
XM_017017276.1 XP_016872765.1 13 - 14 2503 76
XM_017017277.1 XP_016872766.1 12 - 13 1727 76
XM_017017278.1 XP_016872767.1 11 - 12 1727 76
XM_017017279.1 XP_016872768.1 13 - 14 1745 76
XM_017017280.1 XP_016872769.1 13 - 14 2422 76
XM_017017281.1 XP_016872770.1 12 - 13 1922 76
XM_017017282.1 XP_016872771.1 12 - 13 1458 76
XM_017017283.1 XP_016872772.1 12 - 13 1727 76
XM_017017284.1 XP_016872773.1 11 - 12 1303 76
XM_017017285.1 XP_016872774.1 12 - 13 2349 76
XM_017017286.1 XP_016872775.1 9 - 10 1167 76
XM_017017287.1 XP_016872776.1 2 - 3 187 76
GenBank mRNA AB209252.1 - 13 - 14 1790 76
AK126776.1 - 10 - 11 1228 76
AK294375.1 - 12 - 13 1346 76
AK295094.1 - 13 - 14 1815 76
AK295624.1 - 12 - 13 1438 76
AK299797.1 - 13 - 14 1573 76
CR749820.1 - 18 - 19 2139 76
KJ535093.1 - 18 - 19 2022 76
U32376.1 - 13 - 14 1788 76

View Full Product Details