Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG2
Entrez Gene ID
1740
Gene Name
discs large MAGUK scaffold protein 2
Gene Aliases
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location
Chr.11: 83455009 - 85628534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142699.1 NP_001136171.1 14 - 15 1658 84
NM_001206769.1 NP_001193698.1 8 - 9 1342 84
NM_001300983.1 NP_001287912.1 9 - 10 1334 84
NM_001364.3 NP_001355.2 9 - 10 1334 84
XM_005273810.4 XP_005273867.1 8 - 9 977 84
XM_005273811.4 XP_005273868.1 8 - 9 977 84
XM_011544778.2 XP_011543080.1 15 - 16 2949 84
XM_011544780.2 XP_011543082.1 15 - 16 2949 84
XM_011544782.2 XP_011543084.1 10 - 11 1420 84
XM_017017254.1 XP_016872743.1 15 - 16 2949 84
XM_017017255.1 XP_016872744.1 14 - 15 2910 84
XM_017017256.1 XP_016872745.1 14 - 15 2439 84
XM_017017257.1 XP_016872746.1 13 - 14 3371 84
XM_017017258.1 XP_016872747.1 12 - 13 2547 84
XM_017017259.1 XP_016872748.1 13 - 14 2364 84
XM_017017260.1 XP_016872749.1 12 - 13 3296 84
XM_017017262.1 XP_016872751.1 8 - 9 1402 84
XM_017017263.1 XP_016872752.1 10 - 11 1420 84
XM_017017264.1 XP_016872753.1 9 - 10 1597 84
XM_017017265.1 XP_016872754.1 9 - 10 1594 84
XM_017017266.1 XP_016872755.1 8 - 9 1400 84
XM_017017267.1 XP_016872756.1 9 - 10 1502 84
XM_017017268.1 XP_016872757.1 9 - 10 1323 84
XM_017017269.1 XP_016872758.1 9 - 10 2024 84
XM_017017270.1 XP_016872759.1 10 - 11 1420 84
XM_017017271.1 XP_016872760.1 9 - 10 1597 84
XM_017017272.1 XP_016872761.1 9 - 10 1594 84
XM_017017274.1 XP_016872763.1 9 - 10 1502 84
XM_017017276.1 XP_016872765.1 9 - 10 2022 84
XM_017017277.1 XP_016872766.1 8 - 9 1246 84
XM_017017282.1 XP_016872771.1 8 - 9 977 84
XM_017017283.1 XP_016872772.1 8 - 9 1246 84
XM_017017284.1 XP_016872773.1 7 - 8 822 84
XM_017017286.1 XP_016872775.1 5 - 6 686 84
GenBank mRNA AB209252.1 - 9 - 10 1309 84
AK127150.1 - 8 - 9 1340 84
AK294375.1 - 8 - 9 865 84
AK295094.1 - 9 - 10 1334 84
AK295624.1 - 8 - 9 957 84
AK299797.1 - 9 - 10 1092 84
CR749820.1 - 14 - 15 1658 84
KJ535093.1 - 14 - 15 1541 84
U32376.1 - 9 - 10 1307 84

View Full Product Details