Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA1
Entrez Gene ID
2800
Gene Name
golgin A1
Gene Aliases
golgin-97
Chromosome Location
Chr.9: 124878294 - 124941114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002077.3 NP_002068.1 7 - 8 769 69
XM_005251929.3 XP_005251986.1 7 - 8 701 69
XM_006717062.3 XP_006717125.1 7 - 8 765 69
XM_006717063.3 XP_006717126.1 7 - 8 736 69
GenBank mRNA AB208858.1 - 2 - 3 126 69
AK302445.1 - 7 - 8 578 69
AK307789.1 - 4 - 5 436 69
BC032853.1 - 7 - 8 739 69
U51587.1 - 7 - 8 761 69

View Full Product Details