The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018490.2 | NP_060960.2 | 13 - 14 | 1668 | 84 |
GenBank mRNA | AB209743.1 | - | 13 - 14 | 1587 | 84 |
AF257182.1 | - | 13 - 14 | 1627 | 84 | |
BC033039.2 | - | 12 - 13 | 1584 | 84 |